missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gallium ICP Standard, 10,000 ppm Ga in 5% HNO3, Ricca Chemical
$594.60 - $594.60
Chemical Identifiers
| CAS | 7440-55-3 |
|---|---|
| Molecular Formula | Ga |
| Molecular Weight (g/mol) | 69.72 |
| MDL Number | MFCD00134045 |
| InChI Key | GYHNNYVSQQEPJS-UHFFFAOYSA-N |
| PubChem CID | 5360835 |
| ChEBI | CHEBI:49631 |
| IUPAC Name | gallium |
| SMILES | [Ga] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PGA10KN-100
|
Ricca Chemical Company
PGA10KN100 |
100 mL | Natural LDPE Bottle |
Each for $594.60
|
|
||||
Chemical Identifiers
| 7440-55-3 | |
| 69.72 | |
| GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
| CHEBI:49631 | |
| [Ga] |
| Ga | |
| MFCD00134045 | |
| 5360835 | |
| gallium |
Specifications
| Natural LDPE Bottle | |
| 75.0,22.47,0.75 | |
| Liquid | |
| MFCD00134045 | |
| < 2 | |
| GYHNNYVSQQEPJS-UHFFFAOYSA-N | |
| gallium | |
| 100 mL | |
| CHEBI:49631 | |
| Gallium ICP Standard |
Safety and Handling
ShelfLife : 18 months