missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Galacton-Star™ Substrate (50X Concentrate)
Concentrated chemiluminescent substrate for E.coli β-galactosidase assays. Thermo Scientific Galacton-Star Substrate (50X Concentrate) is highly sensitive which makes it ideal for detection of weak expression and for transfection normalization with o
$948.00 - $2,375.00
Specifications
| Product Type | ELISA Substrate |
|---|---|
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Format | Bottle |
| Substrate Type | Beta-Gal |
Description
Galacton-Star substrate is a chemiluminescent substrate designed for the rapid, and sensitive detection of beta-galactosidase in cell lysates. The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric beta-galactosidase assays. Light emission from Galacton-Star reaches maximum in 60-90 minutes and remains constant for at least 1 hour. After incubation at room temperature, the signal is measured in a luminometer. As few as 10 femtograms of beta-galactosidase (20,000 molecules) are detectable.Specifications
| ELISA Substrate | |
| Liquid | |
| Beta-Gal | |
| Room Temperature |
| Chemiluminescence | |
| Bottle | |
| Store the product at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.