Learn More
Invitrogen™ Galacton-Star™ Substrate (50X Concentrate)
Description
Galacton-Star Substrate is a chemiluminescent substrate designed for the rapid, sensitive detection of β-galactosidase in cell lysates. The chemiluminescent assay exhibits over three orders of magnitude greater sensitivity compared to colorimetric β-galactosidase assays. Light emission from Galacton-Star Substrate reaches a maximum in 60-90 minutes and remains constant for at least 1 hour. After incubation at room temperature, the signal is measured in a luminometer.
As few as 10 femtograms of β-galactosidase (20,000 molecules) are detectable. High sensitivity makes this substrate ideal for detection of weak expression and for transfection normalization with other sensitive reporter gene assays.
Specifications
Specifications
| Product Type | ELISA Substrate |
| Quantity | 4 mL |
| Detection Method | Chemiluminescence |
| Form | Liquid |
| Format | Bottle |
| Product Line | Galacton-Star, NovaBright |
| Substrate Type | Beta-Gal |
| Content And Storage | Store the product at 4°C. |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.