missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formalin, 10%, Neutral Buffered, Reagents
Formalin, 10%, Neutral Buffered, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.
$118.04 - $628.89
Specifications
| Chemical Name or Material | Formalin, 10%, Neutral Buffered |
|---|---|
| CAS | 7732-18-5 , 50-00-0 , 67-56-1 , 7558-79-4 , 10049-21-5 |
| Boiling Range | 100°C |
| Color | Colorless |
| Boiling Point | 100°C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-234
|
Labchem Inc
C22085101A |
1 L |
Each for $118.04
|
|
|||||
|
01-337-233
|
Labchem Inc
C220851020F |
20 L |
Each for $628.89
|
|
|||||
|
01-337-232
|
Labchem Inc
C22085104A |
4 L |
Each for $295.11
|
|
|||||
Specifications
| Formalin, 10%, Neutral Buffered | |
| 100°C | |
| 100°C | |
| 1.02 | |
| Reagent |
| 7732-18-5 , 50-00-0 , 67-56-1 , 7558-79-4 , 10049-21-5 | |
| Colorless | |
| HCHO/H2O | |
| Formalin | |
| Liquid |