missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Solution, ACS, 36.5% 38.0%, Spectrum™ Chemical

$234.79 - $2,035.12

Chemical Identifiers

CAS 50-00-0
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
18610650
View Documents
Spectrum Chemical Mfg Cor
F1080125ML
125 mL Poly Drum N/A
Only null left
 
18610651
View Documents
Spectrum Chemical Mfg Cor
F1080500MLPL
500 mL Poly Drum N/A
Only null left
 
18610652
View Documents
Spectrum Chemical Mfg Cor
F10801LT
1 L Poly Drum N/A
Only null left
 
18610653
View Documents
Spectrum Chemical Mfg Cor
F10804LTPL
4 L Poly Drum N/A
Only null left
 
18610648
View Documents
Spectrum Chemical Mfg Cor
F108020LTPL
20 L Poly Drum N/A
Only null left
 
18610649
View Documents
Spectrum Chemical Mfg Cor
F1080200LTBL
200 L Poly Drum N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

50-00-0

Specifications

Liquid
1
0.006 meq/g
125 mL
ACS
50-00-0,7732-18-5,67-56-1
Poly Drum
Colorless
36.5 to 38.0%
Formaldehyde, Solution
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.