missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Solution, ACS, 36.5% 38.0%, Spectrum™ Chemical

Catalog No. 18610650
Change view
Click to view available options
Quantity:
125 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Poly Drum
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18610650 125 mL Poly Drum
18610651 500 mL Poly Drum
18610652 1 L Poly Drum
18610653 4 L Poly Drum
18610648 20 L Poly Drum
18610649 200 L Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18610650 Supplier Spectrum Chemical Mfg Cor Supplier No. F1080125ML
Only null left

Chemical Identifiers

CAS 50-00-0 , 7732-18-5 , 67-56-1

Specifications

Physical Form Liquid
CAS Max % 1
Packaging Poly Drum
Titratable Acid 0.006 meq/g
Color Colorless
Quantity 125 mL
Percent Purity 36.5 to 38.0%
Grade ACS
Chemical Name or Material Formaldehyde, Solution

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.