missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, Solution, ACS, 36.5% 38.0%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor F108020LTPL
Specifications
| Liquid | |
| 50-00-0,7732-18-5,67-56-1 | |
| Poly Drum | |
| Colorless | |
| 36.5 to 38.0% |
| Formaldehyde, Solution | |
| 1 | |
| 0.006 meq/g | |
| 20 L | |
| ACS |
Chemical Identifiers
| 50-00-0 |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.