missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, Solution, ACS, 36.5% 38.0%, Spectrum™ Chemical

Supplier:  Spectrum Chemical Mfg Cor F10801LT

 View more versions of this product

Catalog No. 18610652


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Liquid
50-00-0,7732-18-5,67-56-1
Poly Drum
Colorless
36.5 to 38.0%
Formaldehyde, Solution
1
0.006 meq/g
1 L
ACS

Chemical Identifiers

50-00-0
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.