missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, ACS Reagent Grade, Ricca Chemical

$110.90 - $633.57

Chemical Identifiers

CAS 7732-18-5 , 50-00-0 , 67-56-1
Molecular Formula CH2O
Molecular Weight (g/mol) 30.03
MDL Number MFCD00003274
InChI Key WSFSSNUMVMOOMR-UHFFFAOYSA-N
Synonym formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
PubChem CID 712
ChEBI CHEBI:16842
IUPAC Name formaldehyde
SMILES C=O
View More Specs

Products 6
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
RSOF0010250
View Documents Promotion Details
N/A 250 mL Natural Poly Bottle
Each for $214.23
Only null left
 
RSOF0010500
View Documents Promotion Details
N/A 500 mL Natural Poly Bottle
Each for $110.90
Only null left
 
RSOF00101A
View Documents Promotion Details
N/A 1 L Natural Poly Bottle
Each for $168.10
Only null left
 
RSOF00104A
View Documents Promotion Details
N/A 4 L Natural Poly Bottle
Each for $238.23
Only null left
 
RSOF001010F
View Documents Promotion Details
N/A 10 L Cubitainer
Each for $423.97
Only null left
 
RSOF001020F
View Documents Promotion Details
N/A 20 L Cubitainer
Each for $633.57
Only null left
 
Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 50-00-0 , 67-56-1
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O

Specifications

7732-18-5 , 50-00-0 , 67-56-1
37, 48, 15
1.073
∼96°C
MFCD00003274
Miscible in water
C=O
30.03
CHEBI:16842
ACS
Formaldehyde, 37%, ACS Reagent Grade
37, 48, 15
Colorless
2.8 to 4.0
CH2O
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
37% w/w
Liquid
Videos
Safety and Handling

Safety and Handling

ShelfLife : 60 months

Recommended Storage : Ambient Flammable

SDS
Documents

Documents