missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Formaldehyde, 37%, ACS Reagent Grade, Ricca Chemical
Supplier: Ricca Chemical Company RSOF00101A
| Quantity | 1 L |
|---|---|
| Packaging | Natural Poly Bottle |
Chemical Identifiers
| CH2O | |
| MFCD00003274 | |
| formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
| CHEBI:16842 | |
| C=O |
Specifications
| Formaldehyde | |
| 7732-18-5 , 50-00-0 , 67-56-1 | |
| 38.0, 49.0, 16.0 | |
| Approximately 96°C | |
| CH2O | |
| 1 L | |
| WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
| formaldehyde | |
| 712 | |
| 37% w/w | |
| 37% |
| Aqueous (Formalin) | |
| 37.0, 47.0, 14.0 | |
| Clear colorless | |
| Natural Poly Bottle | |
| MFCD00003274 | |
| formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
| C=O | |
| 30.03 | |
| CHEBI:16842 | |
| ACS | |
| Liquid |
Safety and Handling
ShelfLife : 24 months