missing translation for 'onlineSavingsMsg'
Learn More

Formaldehyde, 37%, ACS Reagent Grade, Ricca Chemical

Supplier:  Ricca Chemical Company RSOF001020F

Quantity 20 L
Packaging Cubitainer

 View more versions of this product

Catalog No. RSOF001020F


Only null left

Description

Description

Specifications

Chemical Identifiers

7732-18-5 , 50-00-0 , 67-56-1
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O

Specifications

Formaldehyde
7732-18-5 , 50-00-0 , 67-56-1
38.0, 49.0, 16.0
Approximately 96°C
CH2O
20 L
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
37% w/w
37%
Aqueous (Formalin)
37.0, 47.0, 14.0
Clear colorless
Cubitainer
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
C=O
30.03
CHEBI:16842
ACS
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

ShelfLife : 24 months

SDS
Documents

Documents

Promotions

Promotions