Learn More
Brady™ Focus LED Line Projectors - Green, Red, Yellow
Focusable projectors deliver more defined, longer lines at increased brightness.
Supplier:  Brady™ 178986
 Afficher plus de versions de ce produit
Numéro de catalogue. 17823777
Il en reste null
Description
Description
  • Mark the unmarkable with Focusable LED Line Projectors.
  • Easily adjust focus for sharp, well-defined projected line edges at different mounting heights.
  • Achieve longer projections without sacrificing output quality.
  • Identify surfaces where paint and tape cannot, including those that are hot, humid, dusty, grimy, uneven and irregular.
  • Eliminate the prep, install, waste and training needed with disposable tapes and paints.
  • Maintenance-free operation with no fans, motors or moving parts.
  • Waterproof design is safe for washdowns.
  • Large heat sink regulates the device in extreme temperatures to ensure long service life.
  • Dimensions (H x W x D): 11.8 x 9.7 x 11.5 in. for 150 W and 10.6 x 10.6 x 17.3 in. for 80 W.
Spécifications
Identifiants chimiques
Gobo Projector
110/240 V
LED
Spécifications
Gobo Projector
110/240 V
LED
150 W
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS