Learn More
Brady™ Focus LED Line Projectors - Green, Red, Yellow
Focusable projectors deliver more defined, longer lines at increased brightness.
Specifications
Product Type Gobo Projector
Voltage 110/240 V
Display LED
View More Specs
Products 2
Catalog Number Mfr. No. Wattage Price Quantity & Availability  
Catalog Number Mfr. No. Wattage Price Quantity & Availability  
17823778
View Documents Promotion Details
Brady™
178987
80 W N/A
Only null left
17823777
View Documents Promotion Details
Brady™
178986
150 W N/A
Only null left
Description
Description
  • Mark the unmarkable with Focusable LED Line Projectors.
  • Easily adjust focus for sharp, well-defined projected line edges at different mounting heights.
  • Achieve longer projections without sacrificing output quality.
  • Identify surfaces where paint and tape cannot, including those that are hot, humid, dusty, grimy, uneven and irregular.
  • Eliminate the prep, install, waste and training needed with disposable tapes and paints.
  • Maintenance-free operation with no fans, motors or moving parts.
  • Waterproof design is safe for washdowns.
  • Large heat sink regulates the device in extreme temperatures to ensure long service life.
  • Dimensions (H x W x D): 11.8 x 9.7 x 11.5 in. for 150 W and 10.6 x 10.6 x 17.3 in. for 80 W.
Specifications
Specifications
Gobo Projector
110/240 V
LED