missing translation for 'onlineSavingsMsg'
Learn More

Fluoride Standard, 0.5 ppm F- (Premixed with TISAB), Ricca Chemical

Catalog No. 3161516
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Natural Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
3161516 500 mL Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3161516 Supplier Ricca Chemical Company Supplier No. 3161.516

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7681-49-4 , 1310-73-2 , 64-19-7 , 7647-14-5 , 13291-61-7 , 7681-49-4
Molecular Formula FNa
Molecular Weight (g/mol) 41.99
MDL Number MFCD00003524
InChI Key PUZPDOWCWNUUKD-UHFFFAOYSA-M
PubChem CID 5235
ChEBI CHEBI:28741
IUPAC Name sodium fluoride
SMILES [F-].[Na+]

Specifications

Packaging Natural Poly Bottle
CAS Min % 93.0, 1.95, 1.64, 1.59, 0.11
CAS Max % 96.0, 2.03, 1.7, 1.65, 0.11
Physical Form Liquid
Color Colorless
Melting Point Approximately 0°C
Quantity 500 mL
Concentration or Composition (by Analyte or Components) 1mL = 0.0005mg F-, 0.5ppm F-, Premixed with TISAB
Chemical Name or Material Fluoride Standard
ShelfLife 12 months