Learn More
BUCHI FlashPure Chromatography Solid Loader
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
$170.82 - $2600.89
Products 6
Catalog Number Mfr. No. Size (Metric) Tube Rack Price Quantity  
Catalog Number Mfr. No. Size (Metric) Tube Rack Price Quantity  
05-402-905
View Documents
BUCHI
148634331
12 g
Pack of 20 Each for $227.25
Only null left
05405537
View Documents
BUCHI
11068975
15 g
Pack of 15 Each for $2,600.89
Only null left
05-402-906
View Documents
BUCHI
148634340
20 g
Pack of 20 Each for $254.70
Only null left
05-402-907
View Documents
BUCHI
148634332
40 g
Pack of 10 Each for $170.82
Only null left
05410123
View Documents
BUCHI
140000157
80 g
Pack of 5 Each for $175.39
Only null left
05410124
View Documents
BUCHI
140000158
120 g
Pack of 5 Each for $202.84
Only null left
Description
Description
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume