Learn More
BUCHI FlashPure Chromatography Solid Loader
Catalog No. 05402905
Change view
Click to view available options
Size (Metric) Tube Rack:
12 g
15 g
20 g
40 g
80 g
120 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size (Metric) Tube Rack
05402905 12 g
05405537 15 g
05402906 20 g
05402907 40 g
05410123 80 g
05410124 120 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 05402905 Supplier BUCHI Supplier No. 148634331
FlashPure Empty Solid Loaders are ideal for samples that are not readily soluble in the desired mobile phase when performing flash chromatography. The sample is adsorbed onto a solid phase (silica or Celite) and then added to the empty solid loader tube.
  • Screw cap makes sealing easy
  • Various sizes for differing sample amounts
  • Choose a size large enough for the sample
  • Insertion Rod allows frit placement and loading of samples smaller than the entire volume
Specifications
Material Polypropylene
Type Empty Solid Loader
Includes Frits
Size (Metric) Tube Rack 12 g