Learn More
Fox West Sales Optical Lens Cleaner
GREENER_CHOICE
Catalog No. 22143974
Change view
Click to view available options
Quantity:
59 mL
Catalog No. Quantity
22143974 59 mL
1 options
Catalog No. 22143974
Edge
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Excellent for removing oil and smudges from objectives or any optical surface
  • Laboratory safe
  • Nonflammable
Specifications
Type Optical Lens Cleaner
Quantity 59 mL