missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Lightweight Cotton Lisle Inspection Gloves

Catalog No. 19013540
Change view
Click to view available options
Size:
Large
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19013540 Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19013540 Supplier Fisherbrand™ Supplier No. ML40
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Ideal for general inspection work, or for use as liners underneath unlined liquid proof gloves.

  • 100% cotton lisle gloves
  • Two-piece construction
  • Wear alone for inspection purposes
  • Can be used as liners under liquid proof gloves
  • Ambidextrous

Specifications

Construction Cut and Sewn
Coating Material Uncoated
Color White
Disposable Single-use
Hand Ambidextrous
Latex Free Yes
Material Cotton
Product Type Inspection Gloves
Size Large
Sterility Non-sterile
For Use With (Application) Inspection, Electronics Assembly
Gender Men's
Length (English) 8.5 in.
Length (Metric) 22 cm
Show More Show Less