Learn More
Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
$14.58 - $15.04
Specifications
Format Roll
Sterility Non-sterile
Quantity 1000/Roll
Diameter (English) 0.37 in.
Diameter (Metric) 9.55 mm
View More Specs
Products 11
Catalog Number Mfr. No. Color Price Quantity & Availability  
Catalog Number Mfr. No. Color Price Quantity & Availability  
11010227
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C24
Blue
Case for $14.95
Only null left
11010230
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C30
Green
Each for $14.95
Only null left
11010228
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C86
Brown
Each for $14.58
Only null left
11010229
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C87
Gold
Each for $14.58
Only null left
11010231
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C88
Lavender
Each for $14.58
Only null left
11010232
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C27
Orange
Each for $15.04
Only null left
11010233
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C28
Pink
Each for $14.95
Only null left
11010234
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C29
Purple
Each for $14.95
Only null left
11010236
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C89
Silver
Each for $14.58
Only null left
11010237
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C25
Yellow
Each for $14.95
Only null left
11010235
GREENER_CHOICE
View Documents Promotion Details
Fisherbrand™
FISH C26
Red
Each for $14.95
Only null left
Description
Description
  • Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Specifications
Specifications
Roll
1000/Roll
9.55 mm
Non-sterile
0.37 in.