Learn More
Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICE
Catalog No. 11010227
Change view
Click to view available options
Color:
Blue
Brown
Gold
Green
Lavender
Orange
Pink
Purple
Red
Silver
Yellow
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
11010227 Blue
11010230 Green
11010228 Brown
11010229 Gold
11010231 Lavender
11010232 Orange
11010233 Pink
11010234 Purple
11010236 Silver
11010237 Yellow
11010235 Red
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Catalog No. 11010227 Supplier Fisherbrand™ Supplier No. FISH C24
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
  • Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Specifications
Color Blue
Format Roll
Product Type Dot Label
Sterility Non-sterile
Quantity 1000/Roll
Diameter (English) 0.37 in.
Diameter (Metric) 9.55 mm