Learn More
Fisherbrand™ 3/8 in. Dot Labels
GREENER_CHOICE
Fisher branded Cryogenic Storage Labels use a special adhesive that is designed to withstand extreme cold temperatures as low as -196°C (-311°F). Our cryo labels can be printed/written on and will not bleed, smudge or smear–perfect for identifying specimens after long periods of cryogenic storage.
Supplier:  Fisherbrand™ FISH C30
 View more versions of this product
Catalog No. 11010230
Only null left
Description
Description
  • Labels can be printed/written on and will not bleed, smudge or smear - perfect for identifying specimens after long periods of cryogenic storage
Specifications
Chemical Identifiers
Roll
1000/Roll
9.55 mm
Non-sterile
0.37 in.
Specifications
Green
Dot Label
1000/Roll
9.55 mm
Roll
Non-sterile
0.37 in.
Product Suggestions
Product Suggestions