missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Fine Pore Cassettes in Tube Packs
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Spécifications
Spécifications
| Pore Style | Fine Pore |
| Quantity | 750/Cs. |
| Closure Included | Yes |
| Closure Type | Attached Lid, Front Hinge |
| PrintMate Compatible | Yes |
| Color | Orange |
| For Use With (Equipment) | For Use With PrintMate™ Printers |
| Product Type | Cassette |