missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Value

Gibco fetal bovine sera offer an excellent value for basic cell culture, specialty research, and specific assays.

Supplier:  Gibco™ A5256501

 Afficher plus de versions de ce produit

Numéro de catalogue. A5256501


Il en reste null

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium

  • Use for general cell culture with common cell lines
  • Low endotoxin and hemoglobin levels
  • Endotoxin level: <10 EU/mL
  • Hemoglobin level: <25 mg/dL
  • Origin: refer to CoA for specific origin. All products may not be available in all regions due to importation regulations.
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco serum delivers

  • ISO 13485–certified; processed in FDA registered facilities
  • Triple-filtered at 0.1 μm
  • Gibco bottle easier to use in hood; helps reduce risk of contamination and improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook

* FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

Spécifications

Identifiants chimiques

Fetal
Fetal Bovine Serum
Triple-filtered, 0.1 μm
Value
Refer to CoA for specific origin*
Sterile
*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.

Spécifications

Storage conditions: ≤-10°C
Fetal
Frozen
Fetal Bovine Serum
Standard (Sterile-filtered)
Triple-filtered, 0.1 μm
Triple filtered at 0.1 μm
≤20 EU/mL
No
Value
Dry Ice
Yes
Cattle/Bovine
Refer to CoA for specific origin*
Bottle
500 mL
Sterile
Value
500 mL
*FBS import requirements vary globally, and acceptance depends on whether competent authorities allow import from a specific country of origin. Please check applicable regulations for the export of FBS and any FBS-containing products (e.g., cultured cells). Contact your Thermo Fisher Scientific representative for support.
≤25 mg/dL
0Y7
USDA Safety Testing Performed for Australian origins
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

missing translation for 'shelfLife' : 5 Years

FDS
Documents

Documents

Promotions

Promotions

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.