missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Nitrate Solution, 404 g/L in dilute Nitric Acid, Ricca Chemical
$643.07 - $1048.40
Chemical Identifiers
| CAS | 7782-61-8 |
|---|---|
| Molecular Formula | FeH18N3O18 |
| Molecular Weight (g/mol) | 403.99 |
| MDL Number | MFCD00149708 |
| InChI Key | SZQUEWJRBJDHSM-UHFFFAOYSA-N |
| Synonym | ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate |
| PubChem CID | 16211566 |
| IUPAC Name | iron(3+) nonahydrate trinitrate |
| SMILES | O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Chemical Identifiers
| 7782-61-8 | |
| 403.99 | |
| SZQUEWJRBJDHSM-UHFFFAOYSA-N | |
| 16211566 | |
| O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
| FeH18N3O18 | |
| MFCD00149708 | |
| ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
| iron(3+) nonahydrate trinitrate |
Specifications
| Yellow | |
| 39.0,64.0,7.0 | |
| FeH18N3O18 | |
| ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate | |
| O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
| 403.99 | |
| Laboratory | |
| Dilute Nitric Acid | |
| 500 mL | |
| 404g/L |