missing translation for 'onlineSavingsMsg'
Learn More

Ferric Nitrate Solution, 404 g/L in dilute Nitric Acid, Ricca Chemical

Catalog No. 313516
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
313516 500 mL Amber Poly Bottle
313532 1 L Amber Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 313516 Supplier Ricca Chemical Company Supplier No. 313516

Chemical Identifiers

CAS 7782-61-8 , 7732-18-5 , 7697-37-2
Molecular Formula FeH18N3O18
Molecular Weight (g/mol) 403.99
MDL Number MFCD00149708
InChI Key SZQUEWJRBJDHSM-UHFFFAOYSA-N
Synonym ferric nitrate nonahydrate, iron iii nitrate nonahydrate, unii-vmx4uop3vn, ferric nitrate, nonahydrate, iron iii nitrate, vmx4uop3vn, iron, reference standard solution, ferricnitratenonahydrate, nitric acid, iron 3+ salt, nonahydrate, iron iii nitrate hydrate
PubChem CID 16211566
IUPAC Name iron(3+) nonahydrate trinitrate
SMILES O.O.O.O.O.O.O.O.O.[Fe+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Color Yellow
CAS Min % 39.0, 64.0, 7.0
CAS Max % 41.0, 67.0, 9.0
Grade Laboratory
Packaging Amber Poly Bottle
Solvent or Matrix Dilute Nitric Acid
pH <1
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 404g/L
Chemical Name or Material Ferric Nitrate Solution
Show More Show Less