missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Solution, Fisher Chemical™

Catalog No. p-213336
Change view
Click to view available options
Quantity:
500 mL
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
SF9000500 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. SF9000500 Supplier Ricca Chemical Company Supplier No. SF9000500
Only null left

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

CAS Min % 0.97, 97.0, 0.24
CAS Max % 1.01, 100.0, 0.25
Percent Purity 1%
Grade Laboratory
Packaging Amber Glass Bottle
Methods For AOAC Method 937.05
Quantity 500 mL
Chemical Name or Material Ferric Chloride Solution
DOTInformation DOT Class 8, : Corrosive