missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride Oxidizing Solution, For Cyanide, Certified, LabChem™

Catalog No. LC143301
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC143301 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC143301 Supplier LabChem Supplier No. LC143301
Only null left

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5 , 5329-14-6
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380  
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Amber to Yellow
CAS Max % 1, 97.4, 1.6
Linear Formula FeCl3·6H2O
Solubility Information Soluble in water
Formula Weight 270.3
Grade Certified
Identification Passes Test
Packaging Poly Bottle
Decomposition Information Hydrogen chloride; Iron oxide; Sulfur compounds
Quantity 500 mL
Sensitivity Passes Test
Physical Form Liquid
Chemical Name or Material Ferric Chloride Oxidizing Solution, For Cyanide
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life.
Precautionary Statement GHS P Statement
Wear protective gloves, eye protection.
Wash exposed skin thoroughly after handling.
Avoid release to the environment.
If on skin: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
Take off contaminated clothing and wash it before reuse.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
Dispose of contents/container to comply with local, state and federal regulations.
Recommended Storage Room Temperature