missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride Oxidizing Solution, 1% (w/v), with 1.6% (w/v) Sulfamic Acid, Ricca Chemical
Specifications
Specifications
| Color | Green-Yellow |
| Percent Purity | 1% |
| Grade | Laboratory |
| Packaging | Natural Poly Bottle |
| Solvent or Matrix | Aqueous,with 1.6% (w/v) Sulfamic Acid |
| pH | Acidic |
| Quantity | 500 mL |
| Physical Form | Liquid |
| Concentration or Composition Notes | 1% (w/v) |
| Chemical Name or Material | Ferric Chloride Oxidizing Solution |