missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 40% (w/v), Ricca Chemical

Catalog No. p-3251675
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
312716 500 mL Natural Poly Bottle
312732 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 312716 Supplier Ricca Chemical Company Supplier No. 312716
Only null left

Chemical Identifiers

CAS 10025-77-1 , 7732-18-5 , 7647-01-0
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Color Amber
CAS Min % 27.78, 69.0, 0.82
CAS Max % 28.91, 72.0, 0.85
Percent Purity 40%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH Acidic
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 40% (w/v)
Chemical Name or Material Ferric Chloride
Show More Show Less