missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ferric Chloride, 0.025% (w/v) as Hexahydrate, for BOD, LabChem™
$58.82 - $83.50
Chemical Identifiers
| CAS | 10025-77-1 |
|---|---|
| Molecular Formula | Cl3FeH12O6 |
| Molecular Weight (g/mol) | 270.29 |
| MDL Number | MFCD00149712 |
| InChI Key | NQXWGWZJXJUMQB-UHFFFAOYSA-K |
| IUPAC Name | iron(3+) hexahydrate trichloride |
| SMILES | O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
Chemical Identifiers
| 10025-77-1 | |
| 270.29 | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| iron(3+) hexahydrate trichloride |
Specifications
| Brown-Yellow | |
| 99.975,0.025 | |
| FeCl3·6H2O | |
| 1g/mL | |
| NQXWGWZJXJUMQB-UHFFFAOYSA-K | |
| iron(3+) hexahydrate trichloride | |
| 270.3 | |
| Poly Bottle | |
| Hydrogen chloride; Iron oxide; Sulfur compounds | |
| Liquid |
| 10025-77-1 , 7732-18-5 | |
| Cl3FeH12O6 | |
| MFCD00149712 | |
| Soluble in water | |
| O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3] | |
| 270.29 | |
| Passes Test | |
| 1g/mL | |
| 500 mL | |
| Ferric Chloride, 0.025% (w/v) as Hexahydrate, for BOD |
Safety and Handling
GHS H Statement
Solution is not hazardous.
GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage : Room Temperature