missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 0.025% (w/v) as Hexahydrate, for BOD, LabChem™

Supplier:  LabChem LC143001

 View more versions of this product

Catalog No. LC143001


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ferric Chloride
0.025% (w/v) as Hexahydrate, for BOD
99.975,0.025
FeCl3·6H2O
1g/mL
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
270.3
Poly Bottle
Hydrogen chloride; Iron oxide; Sulfur compounds
Liquid
Brown-Yellow
10025-77-1 , 7732-18-5
Cl3FeH12O6
MFCD00149712
Soluble in water
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
270.29
Passes Test
1g/mL
500 mL

Chemical Identifiers

10025-77-1
270.29
NQXWGWZJXJUMQB-UHFFFAOYSA-K
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
Cl3FeH12O6
MFCD00149712
iron(3+) hexahydrate trichloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Solution is not hazardous.

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

Recommended Storage : Room Temperature

SDS
Documents

Documents

Promotions

Promotions