missing translation for 'onlineSavingsMsg'
Learn More

Ferric Chloride, 0.025% (w/v), Ricca Chemical

$115.40 - $436.10

Chemical Identifiers

CAS 10025-77-1
Molecular Formula Cl3FeH12O6
Molecular Weight (g/mol) 270.29
MDL Number MFCD00149712
InChI Key NQXWGWZJXJUMQB-UHFFFAOYSA-K
Synonym Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
PubChem CID 24380
ChEBI CHEBI:30808
IUPAC Name iron(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
3100-16
View Documents
Ricca Chemical Company
310016
500 mL Natural Poly Bottle
Each for $115.40
Only null left
 
3100-32
View Documents
Ricca Chemical Company
310032
1 L Natural Poly Bottle
Each for $169.63
Only null left
 
3100-1
View Documents
Ricca Chemical Company
RI31001
4 L Natural Poly Bottle
Each for $436.10
Only null left
 
Description

Description

Specifications

Chemical Identifiers

10025-77-1
270.29
NQXWGWZJXJUMQB-UHFFFAOYSA-K
24380
iron(3+) hexahydrate trichloride
Cl3FeH12O6
MFCD00149712
Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
CHEBI:30808
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]

Specifications

Amber
0.02,98.0,2.0E-4
Cl3FeH12O6
Ferric chloride hexahydrate, Iron Trichloride, Perchlorure de Fer
O.O.O.O.O.O.[Cl-].[Cl-].[Cl-].[Fe+3]
270.29
CHEBI:30808
Laboratory
Aqueous Solution
500 mL
0.025% (w/v)
10025-77-1 , 7732-18-5 , 7647-01-0
0.02,100.0,2.0E-4
MFCD00149712
NQXWGWZJXJUMQB-UHFFFAOYSA-K
iron(3+) hexahydrate trichloride
24380
0.025%
Natural Poly Bottle
Slightly Acidic
Liquid
Ferric Chloride
Videos
SDS
Documents

Documents