missing translation for 'onlineSavingsMsg'
Learn More

Ferric Ammonium Sulfate TS, 8% (w/v), Ricca Chemical

Catalog No. 30584
Change view
Click to view available options
Quantity:
120 mL
500 mL
1 L
Packaging:
Natural Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
30584 120 mL Natural Poly Bottle
305816 500 mL Natural Poly Bottle
305832 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 30584 Supplier Ricca Chemical Company Supplier No. 30584

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7783-83-7 , 7732-18-5
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]

Specifications

Color Amber
CAS Min % 91.0, 7.26
CAS Max % 94.0, 7.56
Percent Purity 8%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH ∼2.5
Quantity 120 mL
Physical Form Liquid
Concentration or Composition Notes 8% (w/v)
Chemical Name or Material Ferric Ammonium Sulfate, Test Solution
Show More Show Less