missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ S1 Nuclease (100 U/μL)

Catalog No. FEREN0321
Change view
Click to view available options
Quantity:
10,000 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEREN0321 10,000 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEREN0321 Supplier Thermo Scientific™ Supplier No. EN0321

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Degrade single-stranded DNA and RNA with this endonuclease.

S1 Nuclease degrades single-stranded nucleic acids, releasing 5'-phosphoryl mono- or oligonucleotides. It is five times more active on DNA than on RNA . S1 Nuclease also cleaves dsDNA at the single-stranded region caused by a nick, gap, mismatch or loop. S1 Nuclease exhibits 3'-phosphomonoesterase activity.

The enzyme is a glycoprotein with carbohydrate content of 18%.

  • Source: Aspergillus oryzae
  • Molecular Weight: 29 kDa monomer
Quality Control:
  • The absence of contaminating double-stranded DNA specific nuclease activity confirmed by appropriate quality test
  • Functionally tested for the generation of unidirectional deletions in DNA fragments (in conjunction with Exonuclease III)
Source:
  • Aspergillus oryzae
Molecular Weight:
  • 29 kDa monomer
Definition of Activity Unit:
  • One unit of the enzyme produces 1μg of acid soluble deoxyribonucleotides in 1 min. at 37°C
  • Enzyme activity is assayed in the following mixture: 30 mM sodium acetate (pH 4.5), 50 mM NaCl, 0.1 mM ZnCl2, 5% (v/v) glycerol, 800μg/ml heat denatured calf thymus DNA
Storage Buffer:
  • The enzyme is supplied in:20mM Tris-HCl (pH 7.5), 50mM NaCl, 0.1mM ZnCl2 and 50% (v/v) glycerol
5X Reaction Buffer:
  • 200mM sodium acetate (pH 4.5 at 25°C), 1.5M NaCl and 10 mM ZnSO4
Inhibition and Inactivation:
  • Inhibitors: metal chelators, PPi, Pi, 5'-ribonucleotides and deoxyribonucleotides
  • Inactivated by heating at 70°C for 10 min. in the presence of EDTA

Recommended for:

Removal of single-stranded overhangs of DNA fragments (2); S1 transcript mapping (3, 4); Cleavage of hairpin loops; Creation of unidirectional deletions in DNA fragments in conjunction with Exonuclease III (5)

Note:

S1 Nuclease can introduce breaks into double-stranded DNA, RNA and DNA/RNA hybrids at high enzyme and low salt concentrations (6).

Specifications

Concentration 100 U/μL
Enzyme Nuclease
Compatible Buffer Storage Buffer, 5X Reaction Buffer
Quantity 10,000 units
Product Type S1 Nuclease

For Research Use Only. Not for use in diagnostic procedures.