missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MreI (Sse232I) (10 U/μL)

Cut at CG^CCGGCG sites with MreI (Sse232I) restriction enzyme, which performs best at 37°C in G buffer.

Supplier:  Thermo Scientific™ ER2021

Numéro de catalogue. FERER2021


Il en reste null

Description

Description

Ad2 DNA digested with MreI (Sse232I), 1% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Spécifications

Spécifications

10 U/μL
MreI (Sse232I)
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer G
37°C
300 units
Traditional Cloning
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.