missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MluI (10 U/μL)

The MluI restriction enzyme recognizes A^CGCGT sites and cuts best at 37°C in R buffer.

Supplier:  Thermo Scientific™ ER0562

 Afficher plus de versions de ce produit

Numéro de catalogue. FERER0562


Il en reste null

Description

Description

Lambda DNA digested with MluI, 0.7% agarose, 7 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

5'...A▵C G C G T...3'

3'...T G C G C▵A...5'

Conditions for 100% Activity:

  • 1X Buffer R: 10mM Tris-HCl (pH 8.5 at 37°C), 10mM MgCl2, 100mM KCl and 0.1mg/mL BSA
  • Incubate at 37°C

Storage Buffer:

  • MluI is supplied in: 10mM Tris-HCl (pH 7.5 at 25°C), 50mM KCl, 1mM DTT, 0.1mM EDTA, 0.2mg/mL BSA and 50% (v/v) glycerol

Methylation Effects:

  • Dam: never overlaps —no effect
  • Dcm: never overlaps —no effect
  • CpG: completely overlaps —blocked
  • EcoKI: may overlap —no effect
  • EcoBI: may overlap —no effect

Digestion of Agarose-embedded DNA:

  • Minimum 5U of the enzyme are required for complete digestion of 1μg of agarose-embedded lambda DNA in 16 hours

Compatible Ends:

  • AflIII, BtgI, PauI, SgsI
Spécifications

Identifiants chimiques

10 U/μL
Mlu I
Yes
No
Traditional Cloning
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer R
37°C
Restriction Enzyme

Spécifications

10 U/μL
Mlu I
Yes
No
Restriction Enzyme
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer R
37°C
5,000 units
Traditional Cloning
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
FDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.