missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ LweI (SfaNI) (10 U/μL)

The LweI (SfaNI) restriction enzyme recognizes GCATC(5/9)^ sites and cuts best at 37°C in Tango buffer (Isoschizomers: SfaNI).

Spécifications

Concentration 10 U/μL
Methylation Sensitivity CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
Enzyme LweI (SfaNI)
Compatible Buffer 10X Buffer Tango
Sensitive to Heat Inactivation Yes
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
FERER1621
Afficher les documents Détails de la promotion
Thermo Scientific™
ER1621
100 units N/A
Il en reste null
 
FERER1622
Afficher les documents Détails de la promotion
Thermo Scientific™
ER1622
500 units N/A
Il en reste null
 
Description

Description

Lambda DNA digested with LweI (SfaNI), 1.4% agarose, 169 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Spécifications

Spécifications

10 U/μL
LweI (SfaNI)
Yes
Yes
Scarless Cloning
CpG Methylation-Sensitive, Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X Buffer Tango
37°C
Restriction Enzyme
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.