missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BamHI-Lsp1109I Buffer (10X)

Numéro de catalogue. FERB57
Modifier l'affichage
Click to view available options
Quantity:
5 x 1.0 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity
FERB57 5 x 1.0 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Numéro de catalogue. FERB57 Fournisseur Thermo Scientific™ Code fournisseur B57
Il en reste null

Optimize digestions with BamHI and Lsp1109I restriction enzymes with this buffer, premixed with BSA for enhanced stability.

To ensure consistent enzyme performance, Thermo Scientific restriction enzyme buffers contain BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations. Multiple freeze-thaw cycles of the buffers will not cause BSA precipitation.

restriction enzymes exhibit 100% of their certified activity in the recommended buffer.

The following enzymes require unique buffers for optimal digestion: 

  • AarI 
  • AjiI 
  • BamHI 
  • BfuI 
  • Bpu10I 
  • BseXI 
  • Bsp143I 
  • Cfr9I 
  • Cfr10I 
  • Eam1105I 
  • Ecl136II 
  • Eco52I 
  • EcoRI 
  • KpnI 
  • Lsp1109I 
  • PacI 
  • PasI 
  • Ppu21I 
  • SacI 
  • ScaI 
  • SdaI 
  • SduI 
  • SgeI 
  • TaqI 

Spécifications

Concentration 10X
Quantity 5 x 1.0 mL
Product Type BamHI-Lsp1109I Buffer
Research Category Traditional Cloning

For Research Use Only. Not for use in diagnostic procedures.