missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Blue No. 2, Spectrum™ Chemical
FD115, 860-22-0, C16H8N2O8S2Na2
$348.29 - $26,720.75
Chemical Identifiers
| CAS | 860-22-0 |
|---|---|
| Molecular Formula | C16H8N2Na2O8S2 |
| Molecular Weight (g/mol) | 466.35 |
| MDL Number | MFCD00005723 |
| InChI Key | KHLVKKOJDHCJMG-QDBORUFSSA-L |
| IUPAC Name | disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate |
| SMILES | [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
18602611
|
N/A | 25 g | Amber Glass Bottle |
Each for $348.29
|
|
||||
|
18602612
|
N/A | 100 g | Amber Glass Bottle |
Each for $702.25
|
|
||||
|
18602613
|
N/A | 500 g | Amber Glass Bottle |
Each for $1,777.25
|
|
||||
|
18602615
|
N/A | 5 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18602614
|
N/A | 10 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical FD and C Blue No. 2Chemical Identifiers
| 860-22-0 | |
| 466.35 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| C16H8N2Na2O8S2 | |
| MFCD00005723 | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate |
Specifications
| 860-22-0 | |
| C16H8N2Na2O8S2 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate | |
| 85% | |
| Dark Blue |
| 100% | |
| MFCD00005723 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O | |
| 466.35 | |
| Reagent | |
| FD and C Blue No. 2 |