missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Blue No. 2, Spectrum™ Chemical
Description
Spectrum™ Chemical FD and C Blue No. 2
Specifications
Specifications
| CAS Max % | 100% |
| Packaging | Amber Glass Bottle |
| Color | Dark Blue |
| Quantity | 5 kg |
| Percent Purity | 85% |
| Grade | Reagent |
| Chemical Name or Material | FD and C Blue No. 2 |