missing translation for 'onlineSavingsMsg'
Learn More

FD and C Blue No. 2, Spectrum™ Chemical

FD115, 860-22-0, C16H8N2O8S2Na2

Supplier:  Spectrum Chemical Mfg Cor FD115500GM

Quantity 500 g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. 18602613


Only null left

Description

Description

Spectrum™ Chemical FD and C Blue No. 2
Specifications

Chemical Identifiers

860-22-0
466.35
KHLVKKOJDHCJMG-QDBORUFSSA-L
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
C16H8N2Na2O8S2
MFCD00005723
disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate

Specifications

FD and C Blue No. 2
100%
MFCD00005723
[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O
466.35
Reagent
Dark Blue
860-22-0
C16H8N2Na2O8S2
KHLVKKOJDHCJMG-QDBORUFSSA-L
disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
85%
Amber Glass Bottle
500 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions