missing translation for 'onlineSavingsMsg'
Learn More
Learn More
FD and C Blue No. 2, Spectrum™ Chemical
FD115, 860-22-0, C16H8N2O8S2Na2
Supplier: Spectrum Chemical Mfg Cor FD11510KGBL
| Quantity | 10 kg |
|---|---|
| Packaging | Amber Glass Bottle |
Description
Spectrum™ Chemical FD and C Blue No. 2Chemical Identifiers
| 860-22-0 | |
| 466.35 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O |
| C16H8N2Na2O8S2 | |
| MFCD00005723 | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate |
Specifications
| FD and C Blue No. 2 | |
| 100% | |
| MFCD00005723 | |
| [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O | |
| 466.35 | |
| Reagent | |
| Dark Blue |
| 860-22-0 | |
| C16H8N2Na2O8S2 | |
| KHLVKKOJDHCJMG-QDBORUFSSA-L | |
| disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate | |
| 85% | |
| Amber Glass Bottle | |
| 10 kg |