missing translation for 'onlineSavingsMsg'
Learn More

FD and C Blue No. 2, Spectrum™ Chemical

Catalog No. 18602611
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
5 kg
10 kg
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602611 25 g Amber Glass Bottle
18602612 100 g Amber Glass Bottle
18602613 500 g Amber Glass Bottle
18602615 5 kg Amber Glass Bottle
18602614 10 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602611 Supplier Spectrum Chemical Mfg Cor Supplier No. FD11525GM
Get Up to $500 in Chemicals with Purchase
View all promotions

FD115, 860-22-0, C16H8N2O8S2Na2

Spectrum™ Chemical FD and C Blue No. 2

Chemical Identifiers

CAS 860-22-0
Molecular Formula C16H8N2Na2O8S2
Molecular Weight (g/mol) 466.35
MDL Number MFCD00005723
InChI Key KHLVKKOJDHCJMG-QDBORUFSSA-L
IUPAC Name disodium (E)-3,3'-dioxo-1H,1'H,3H,3'H-[2,2'-biindolylidene]-5,5'-disulfonate
SMILES [Na+].[Na+].[O-]S(=O)(=O)C1=CC=C2N\C(C(=O)C2=C1)=C1\NC2=CC=C(C=C2C1=O)S([O-])(=O)=O

Specifications

CAS Max % 100%
Percent Purity 85%
Grade Reagent
Packaging Amber Glass Bottle
Color Dark Blue
Quantity 25 g
Chemical Name or Material FD and C Blue No. 2