missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Favipiravir

CAS: 259793-96-9 | C5H4FN3O2, C5H4FN3O2 | 157.10 g/mol

$362.02 - $970.75

Chemical Identifiers

CAS 259793-96-9
Molecular Formula C5H4FN3O2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC468691000
View Documents
Thermo Scientific Chemicals
468691000
100 mg
Each for $362.02
Only null left
 
AC468695000
View Documents
Thermo Scientific Chemicals
468695000
500 mg
Each for $970.75
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

259793-96-9
C5H4FN3O2

Specifications

259793-96-9
52.6°C
157.09
NC(=O)C1=NC(F)=CNC1=O
157.10
100 mg
C5H4FN3O2
ZCGNOVWYSGBHAU-UHFFFAOYSA-N
6-fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide
Favipiravir
Videos
Safety and Handling

Safety and Handling

GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only