missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Favipiravir

Catalog No. ac468695000
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 mg
500 mg
Catalog No. Quantity
AC468695000 500 mg
AC468691000 100 mg
2 options

Catalog No. AC468695000

Supplier: Thermo Scientific Chemicals 468695000

Only null left

CAS: 259793-96-9 | C5H4FN3O2, C5H4FN3O2 | 157.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 259793-96-9
Molecular Formula C5H4FN3O2

Specifications

Quantity 500 mg
Boiling Point 52.6°C
Formula Weight 157.09
InChI Key ZCGNOVWYSGBHAU-UHFFFAOYSA-N
SMILES NC(=O)C1=NC(F)=CNC1=O
IUPAC Name 6-fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide
Molecular Weight (g/mol) 157.10
Chemical Name or Material Favipiravir
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only