missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Favipiravir

Catalog No. ac468691000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC468691000 100 mg
AC468695000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC468691000 Supplier Thermo Scientific Chemicals Supplier No. 468691000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 259793-96-9 | C5H4FN3O2, C5H4FN3O2 | 157.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 259793-96-9
Molecular Formula C5H4FN3O2

Specifications

Quantity 100 mg
Boiling Point 52.6°C
Formula Weight 157.09
InChI Key ZCGNOVWYSGBHAU-UHFFFAOYSA-N
SMILES NC(=O)C1=NC(F)=CNC1=O
IUPAC Name 6-fluoro-3-oxo-3,4-dihydropyrazine-2-carboxamide
Molecular Weight (g/mol) 157.10
Chemical Name or Material Favipiravir
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Causes skin irritation.

Causes serious eye irritation.

May cause respiratory irritation.

Precautionary Statement GHS P Statement IF ON SKIN: Wash with plenty of soap and water.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only