missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest DpnI

Cut at Gm6A^TC sites with Thermo Scientific™ FastDigest™ DpnI, which performs best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: MalI).

$55.06 - $106.16

Spécifications

Methylation Sensitivity Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
Enzyme Dpn I
Compatible Buffer 10X FastDigest Buffer/FastDigest Green Buffer
Sensitive to Heat Inactivation Yes
Optimal Reaction Temperature 37°C
voir les spécifications

Produits 2
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
Numéro de catalogue Numéro du manufacturier. Quantity Prix Quantity & Availability  
FERFD1703
Afficher les documents Détails de la promotion
Thermo Scientific™
FD1703
50 μL (50 Reactions)
chaque for $55.06
Il en reste null
 
FERFD1704
Afficher les documents Détails de la promotion
Thermo Scientific™
FD1704
100 μL (100 Reactions)
chaque for $106.16
Il en reste null
 
Description

Description

Lambda DNA digested with DpnI, 1.4% agarose, 22 cleavage sites

FastDigest enzymes are an advanced line of fast restriction enzymes that are all 100% active in the universal FastDigest and FastDigest Green reaction buffers.

The universal buffer allows rapid single-, double- or multiple DNA digestion within 5-15 minutes eliminating any need for buffer change or subsequent DNA clean-up steps. DNA modifying enzymes, such as Klenow Fragment, T4 DNA Ligase, alkaline phosphatases and T4 DNA Polymerase all have 100% activity in FastDigest Buffer. Therefore, enzymes for downstream applications can be directly added to the FastDigest reaction mix.

For additional convenience FastDigest Green Buffer includes a density reagent and two tracking dyes for direct loading of digestion reaction products on gels.

Short incubation times and optimal composition of the universal FastDigest Buffer eliminate star activity effects.

Highlights

  • 100% activity of all FastDigest enzymes in the universal buffer
  • 100% buffer compatibility with downstream applications
  • Complete digestion in 5-15 minutes
  • Direct loading on gels
  • No star activity
  • 176 FastDigest enzymes available
Spécifications

Spécifications

Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
Restriction Enzyme
FastDigest
80°C
Dpn I
Yes
No
Traditional Cloning
Blunt
Videos
FDS
Documents

Documents

For Research Use Only. Not for use in diagnostic procedures.