missing translation for 'onlineSavingsMsg'
Learn More

Dentec Safety FaceTec™ Double Crown Faceshield

Catalog No. 296351013
Change view
Click to view available options
Product Type:
Full Coverage Faceshield
Style:
Capmount
Lens Type:
None
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Style Lens Type
296351013 Full Coverage Faceshield Capmount None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 296351013 Supplier Dentec Safety Supplier No. 12F483CAPO

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

FaceTec™ Double Crown Faceshield provides comfortable, complete coverage for the face and neck.

  • Molded from high strength polycarbonate material
  • Made to withstand rugged use
  • Comfortable fit

Specifications

Shape Rectangle
Lens Coating Uncoated
Lens Type None
Height (English) 12 in.
Width (English) 9 in.
Width (Metric) 22.86 cm
Height (Metric) 30.48 cm
Lens Tint None
Product Type Full Coverage Faceshield
Style Capmount
Material Polycarbonate
Disposable Reusable
Lens Material None
Frame Material Polycarbonate
UV Protection None
Show More Show Less