missing translation for 'onlineSavingsMsg'
Learn More

EDTA disodium salt solution with zinc complex added (solution B), 1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry, Honeywell Fluka™

1 ml solution = 1 German degree of hardness in 100 ml of water, for complexometry

$219.32 - $219.32

Specifications

CAS 7646-85-7 , 60-00-4 , 7732-18-5
UN Number NONH for all modes of transport
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
6002528
View Documents
Honeywell Chemicals
345441L
1 L Poly Bottle
Each for $219.32
Only null left
 
Description

Description

Honeywell´s high-quality titration products cover the complete titration workflow and include:

  • Solutions for volumetric titration (acids, bases, or salts)
  • Buffers available as concentrates or ready-to-use

 

  • Reagents for complexometry:
    • Aminopolycarboxylic acids (EDTA/NTA analogs)
    • Masking agents
  • Indicators for volumetric and complexometric titrations

 


 

Specifications

Specifications

7646-85-7,60-00-4,7732-18-5
NONH for all modes of transport
Videos
SDS
Documents

Documents