missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethylenediaminetetraacetic Acid (0.5M Solution/pH 8.0), Fisher BioReagents
$116.75 - $1,378.47
Chemical Identifiers
| CAS | 60-00-4 |
|---|---|
| Molecular Formula | C10H16N2O8 |
| Molecular Weight (g/mol) | 292.24 |
| MDL Number | MFCD00003541 |
| InChI Key | KCXVZYZYPLLWCC-UHFFFAOYSA-N |
| Synonym | edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol |
| PubChem CID | 6049 |
| ChEBI | CHEBI:42191 |
| IUPAC Name | 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
| SMILES | OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
BP2482100
|
Fisher BioReagents
BP2482100 |
100 mL | Poly Bottle |
N/A
|
|
||||
|
BP2482500
|
Fisher BioReagents
BP2482500 |
500 mL | Poly Bottle |
N/A
|
|
||||
|
BP24821
|
Fisher BioReagents
BP24821 |
1 L | Nalgene Poly Bottle |
N/A
|
|
||||
|
BP248220
|
Fisher BioReagents
BP248220 |
20 L | Poly CUBE |
N/A
|
|
||||
Description
EDTA is used extensively in molecular biology to minimize metal ion impurities in reaction buffers. Ideal for DNA work.Chemical Identifiers
| 60-00-4 | |
| 292.24 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 6049 | |
| 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid |
| C10H16N2O8 | |
| MFCD00003541 | |
| edta, edetic acid, ethylenediaminetetraacetic acid, edathamil, versene, endrate, havidote, titriplex, edta acid, sequestrol | |
| CHEBI:42191 | |
| OC(=O)CN(CCN(CC(O)=O)CC(O)=O)CC(O)=O |
Specifications
| Colorless | |
| C10H16N2O8 | |
| MFCD00003541 | |
| 15, 3565 | |
| KCXVZYZYPLLWCC-UHFFFAOYSA-N | |
| 2-[2-[bis(carboxymethyl)amino]ethyl-(carboxymethyl)amino]acetic acid | |
| 6049 | |
| 292.25 | |
| Poly Bottle | |
| 100 mL | |
| Ethylenediamine Tetraacetic Acid, 0.5 M Solution, pH 8.0 (for DNA Work) |
Safety and Handling
Recommended Storage : RT